Learn More
Description
- Provides barrier between the floor and the bottom shelf contents to deter contamination.
- Features a 1/8 in. raised ship's edge on all four sides to contain spills.
- Type 304 stainless steel construction with epoxy-coated cast corners address the majority of applications for solid, corrosion-resistant shelves or work surfaces.
Specifications
Specifications
| Product Type | Shelf |
| Color | Black, Silver |
| Material | Stainless Steel |
| Autoclavable | Not Autoclavable |
| Certifications/Compliance | NSF Listed |
| Dimensions (L x W) | 18 x 24 in. |
| For Use With (Application) | Stainless Steel shelving |
| For Use With (Equipment) | Metro Super Erecta Shelving |
| Includes | Stainless Steel Shelf and Split Sleeves |
| Warranty | Product will be free from defects in material and workmanship under normal use, service and maintenance for a period of one (1) year from the date of shipment of the Product by Seller, unless otherwise specified by Seller in writing. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.