Learn More
Bellco Glass, Inc. 182cm2 Tissue Culture Flask
SDP
Catalog No. 502329916
Encompass_Preferred
Tissue Culture Flasks are available in Tissue Culture Treated and Nontreated Suspension Culture Versions Tissue Culture Treated Flasks have uniform hydrophilic surface ideal for consistent cell attachment Convenient zipclosure bag is a practical alternative to folding and taping open bags Large opening canted neck design and moderate angles allow easy access to entire flask surface ideal for cell scrapers Stable base enables filling with flask standing upright Thick durable walls Flask is printed with lot number and expiration date for easy identification and tracking Molded interlocking T design for improved stacking on 75 182 amp 300cm2 and 250 600 amp 850mL size flasks Vent caps feature 0.22µm hydrophobic membraneSpecifications Size 182cm Pack Description 5/Resealable Bag Max Volume
Catalog No. 50-232-9916 Supplier Bellco Glass, Inc. Supplier No. 229350000
Tissue Culture Flasks are available in Tissue Culture Treated and Nontreated Suspension Culture Versions Tissue Culture Treated Flasks have uniform hydrophilic surface ideal for consistent cell attachment Convenient zipclosure bag is a practical alternative to folding and taping open bags Large opening canted neck design and moderate angles allow easy access to entire flask surface ideal for cell scrapers Stable base enables filling with flask standing upright Thick durable walls Flask is printed with lot number and expiration date for easy identification and tracking Molded interlocking T design for improved stacking on 75 182 amp 300cm2 and 250 600 amp 850mL size flasks Vent caps feature 0.22µm hydrophobic membraneSpecifications Size 182cm Pack Description 5/Resealable Bag Max Volume