Learn More
eMolecules​ 183673-71-4 | 1-Boc-4-Amino-piperidine-4-carboxylic acid | Chemcia | MFCD01318728 | 244.291 | C11H20N2O4 | 97.000 | CC(C)(C)OC(=O)N1CCC(N)(CC1)C(O)=O | 5g | 357554964
Catalog No. 5000567141 Shop All eMolecules Inc. Products
Encompass_Preferred
1-Boc-4-Amino-piperidine-4-carboxylic acid | Chemcia | 183673-71-4 | MFCD01318728 | 244.291 | C11H20N2O4 | 97.000 | CC(C)(C)OC(=O)N1CCC(N)(CC1)C(O)=O | 5g | 357554964
Catalog No. 50-005-67141 Supplier eMolecules​ Supplier No. 357554964
1-Boc-4-Amino-piperidine-4-carboxylic acid | Chemcia | 183673-71-4 | MFCD01318728 | 244.291 | C11H20N2O4 | 97.000 | CC(C)(C)OC(=O)N1CCC(N)(CC1)C(O)=O | 5g | 357554964