Learn More
ATCC 184A1; Breast Epithelial Cells; Human (Homo sapiens)
Supplier:  ATCC CRL8798
Encompass
184A1 is an epithelial-like cell was isolated from the normal mammary tissue of a 21-year-old female from a reduction mammoplasty. Cells were exposed to benzopyrene and the transformed cell line was established.
Catalog No. 50-238-2836