Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 18Alpha-Glycyrrhetinic Acid

Catalog No. G85031G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G85031G 1 g
G8503250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. G85031G Supplier Sigma Aldrich Supplier No. G85031G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

18+a-Glycyrrhetinic acid (18+a-GA) is a bioactive triterpenoid found in licorice. It shows selective inhibition of 11-HSD1 (11-hydroxysteroid dehydrogenase 1). It also shows anti-proliferative and apoptotic effect on hepatic stellate cells (HSCs).18+a-Glycyrrhetinic acid may be used to synthesize:3-keto-18+a-glycyrrhetinic acid methyl esters of 18+a-glycyrrhetinic acidmethyl esters of 3-keto-18+a-glycyrrhetinic acid

Specifications

CAS 51166-71-3
Molecular Formula C56H98O35
Linear Formula C56H98O35
MDL Number MFCD00011616
Quantity 1 g
Synonym 2,6-Di-O-methyl-beta-cyclodextrin; Dimethyl beta-cyclodextrin
Molecular Weight (g/mol) 1331.36 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.