Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 18Alpha-Glycyrrhetinic Acid

Catalog No. G8503250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G8503250MG 250 mg
G85031G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. G8503250MG Supplier Sigma Aldrich Supplier No. G8503250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

18+a-Glycyrrhetinic acid (18+a-GA) is a bioactive triterpenoid found in licorice. It shows selective inhibition of 11-HSD1 (11-hydroxysteroid dehydrogenase 1). It also shows anti-proliferative and apoptotic effect on hepatic stellate cells (HSCs).18+a-Glycyrrhetinic acid may be used to synthesize:3-keto-18+a-glycyrrhetinic acid methyl esters of 18+a-glycyrrhetinic acidmethyl esters of 3-keto-18+a-glycyrrhetinic acid

Specifications

CAS 1449-05-4
Molecular Formula C30H46O4
Linear Formula C30H46O4
MDL Number MFCD00064897
Quantity 250 mg
Synonym 18alpha-Glycyrrhetic acid; 18-Isoglycyrrhetinic acid; 3beta-Hydroxy-11-oxo-18alpha,20beta-olean-12-en-29-oic acid
Molecular Weight (g/mol) 470.68 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.