Learn More
Ace Glass, Inc. 18g 152mm stainless steel needles, cs/12, sp/6
SDP
Catalog No. 509559111
Encompass
18 gauge 152mm stainless steel needle with a deflected septum point and a standard female hub. Case of 12.
Catalog No. 50-955-9111 Supplier Ace Glass, Inc. Supplier No. 1368415
18 gauge 152mm stainless steel needle with a deflected septum point and a standard female hub. Case of 12.