Learn More
Fine Science Tools Reusable Feeding Needles, 18 Gauge, 38 mm
SDP
Catalog No. NC1649998
Encompass
Product: Reusable Feeding Needles - Straight/38mm/18G/2.25mm Tip Diameter/ Gauge: 18/ Tip Diameter: 2.25mm/ Shape: Straight/ Tip Shape: Round/ Material: Stainless Steel/ Length: 38mm/ Sterile: No/ Style: Reusable/ Unit
Catalog No. NC1649998 Supplier Fine Science Tools Supplier No. 1806138
Product: Reusable Feeding Needles - Straight/38mm/18G/2.25mm Tip Diameter/ Gauge: 18/ Tip Diameter: 2.25mm/ Shape: Straight/ Tip Shape: Round/ Material: Stainless Steel/ Length: 38mm/ Sterile: No/ Style: Reusable/ Unit