Learn More
Cayman Chemical 18-hydroxy Oleoyl Leucine-d7, Purity: ≥99% deuterated forms (d1-d7), 1 mg
SDP
Catalog No. 502895935
Encompass_Preferred
An internal standard for the quantification of 18-hydroxy oleoyl leucine by GC- or LC-MS
Catalog No. 50-289-5935 Supplier Cayman Chemical Supplier No. 90043311 MG
An internal standard for the quantification of 18-hydroxy oleoyl leucine by GC- or LC-MS