Learn More
Cayman Chemical 18hydroxy Oleoyl LeucInd7 50UG
SDP
Catalog No. 502895311
Encompass_Preferred
An internal standard for the quantification of 18-hydroxy oleoyl leucine by GC- or LC-MS
Catalog No. 50-289-5311 Supplier Cayman Chemical Supplier No. 900433150 UG
An internal standard for the quantification of 18-hydroxy oleoyl leucine by GC- or LC-MS