Learn More
Analytical Sales & Services 18mm, 10mL, 22.5 x 46mm Clear Glass Screw Thread Head Space Vials
Catalog No. 5000806090
Encompass
Advantage™ 18mm, 10mL Volume, 22.5 x 46mm Clear Glass Screw Thread Head Space Vials Flat Bottom
Catalog No. 50-008-06090 Supplier Analytical Sales & Services Supplier No. 62546
Advantage™ 18mm, 10mL Volume, 22.5 x 46mm Clear Glass Screw Thread Head Space Vials Flat Bottom