Learn More
Beckman Coulter Adapter, 1/4-28 to M6
Catalog No. NC2244059
Encompass_Preferred
The Beckman Coulter Microfuge 18 Centrifuge Rotor, designed for the Microfuge 18 series, has a 6-place capacity for 1.5/2.0mL tubes and achieves up to 18,213 x g. Its durable polycarbonate build and aerosol-resistant lid make it ideal for rapid pelleting in molecular biology and chemistry applications.
This item is not returnable. View return policy
Catalog No. NC2244059 Supplier Beckman Coulter Supplier No. 359473
The Beckman Coulter Microfuge 18 Centrifuge Rotor, designed for the Microfuge 18 series, has a 6-place capacity for 1.5/2.0mL tubes and achieves up to 18,213 x g. Its durable polycarbonate build and aerosol-resistant lid make it ideal for rapid pelleting in molecular biology and chemistry applications.