Learn More
Beckman Coulter Adapter, Sample Cup, 13 x 75 mm, Polypropylene, Sterile, Pack of 500
Catalog No. NC1712907
Encompass_Preferred
The Beckman Coulter Microfuge 18 Centrifuge Rotor Adapter is designed to adapt 0.2mL and 0.5mL microcentrifuge tubes to fit in the Microfuge 18 centrifuge rotor. It allows the centrifuge to spin smaller volume tubes that normally would not fit. The adapter is made of durable polypropylene and can hold up to 18 microcentrifuge tubes. It has an inner diameter of 11mm and outer diameter of 18mm. The adapter should only be used in the Microfuge 18 centrifuge up to 15,000 rpm. It enables the versatile use of the Microfuge 18 for small volume samples.
Catalog No. NC1712907 Supplier Beckman Coulter Supplier No. 359152
The Beckman Coulter Microfuge 18 Centrifuge Rotor Adapter is designed to adapt 0.2mL and 0.5mL microcentrifuge tubes to fit in the Microfuge 18 centrifuge rotor. It allows the centrifuge to spin smaller volume tubes that normally would not fit. The adapter is made of durable polypropylene and can hold up to 18 microcentrifuge tubes. It has an inner diameter of 11mm and outer diameter of 18mm. The adapter should only be used in the Microfuge 18 centrifuge up to 15,000 rpm. It enables the versatile use of the Microfuge 18 for small volume samples.