Learn More
Hampton Research 18 mm Mounted CryoLoop? - 20 micron.
SDP
Catalog No. NC0375431
Encompass
18 mm Mounted CryoLoop? - 20 micron. Ready to use 18 mm length MicroTube with CryoLoop -Synthetic CryoLoop on stainless steel MicroTube -Nylon loops are staked to hollow, stainless steel MicroTubes? that are used to mount, freeze, and secure the crystal during cryocrystallographic procedures and X-ray data collection. The MicroTube on the 18 mm Mounted CryoLoops is 18 mm in length and ready to insert into CrystalCap Magnetic and CrystalCap HT. The 20 micron diameter nylon loops show minimal diffraction, are thin for fast cryo cooling of the crystal, strong, and aerodynamic.. Please note items from this supplier are non-returnable.
Catalog No. NC0375431 Supplier Hampton Research Supplier No. HR4970
18 mm Mounted CryoLoop? - 20 micron. Ready to use 18 mm length MicroTube with CryoLoop -Synthetic CryoLoop on stainless steel MicroTube -Nylon loops are staked to hollow, stainless steel MicroTubes? that are used to mount, freeze, and secure the crystal during cryocrystallographic procedures and X-ray data collection. The MicroTube on the 18 mm Mounted CryoLoops is 18 mm in length and ready to insert into CrystalCap Magnetic and CrystalCap HT. The 20 micron diameter nylon loops show minimal diffraction, are thin for fast cryo cooling of the crystal, strong, and aerodynamic.. Please note items from this supplier are non-returnable.