Learn More
Molecular Dimensions 18MM ROUND COVER SLIPS (1000)
SDP
Supplier:  Molecular Dimensions MD407
Encompass
18mm round cover slips (1000) Non-Siliconized. No.2 size = 0.19 mm - 0.23 mm thickness- standard for crystallization.
Catalog No. 50-103-8555