Learn More
Molecular Dimensions 18MM ROUND COVER SLIPS (1000)
SDP
Catalog No. 501038552
Encompass
18mm round cover slips (1000) Siliconized. No.2 size = 0.19 mm - 0.23 mm thickness- standard for crystallization.UV compatible
Catalog No. 50-103-8552 Supplier Molecular Dimensions Supplier No. MD402
18mm round cover slips (1000) Siliconized. No.2 size = 0.19 mm - 0.23 mm thickness- standard for crystallization.UV compatible