Learn More
Uline 18 x 18 x 36 Tall Corrugated Boxes | N/A | 1 EA
Catalog No. NC1845845
Encompass_Preferred
Eliminate custom box orders with Ulines stock tall shipping boxes, Ship irregularly shaped items like golf clubs lamps carpet rolls and fishing poles, Tall shipping boxes open on ends, Requires less tape than long boxes, 200 LB TEST, SOLD IN BUNDLE QUANTITIES, All boxes open along the length first dimension, Seams are glued, Meets ASTM D5118, 200 test boxes withstand 200 lbssqin of side wall pressure before bursting
Catalog No. NC1845845 Supplier Uline Supplier No. S4638
Eliminate custom box orders with Ulines stock tall shipping boxes, Ship irregularly shaped items like golf clubs lamps carpet rolls and fishing poles, Tall shipping boxes open on ends, Requires less tape than long boxes, 200 LB TEST, SOLD IN BUNDLE QUANTITIES, All boxes open along the length first dimension, Seams are glued, Meets ASTM D5118, 200 test boxes withstand 200 lbssqin of side wall pressure before bursting