Learn More
MSC PRO-SOURCE Pleated Air Filter, MERV 10, Synthetic, 1 ea
Catalog No. 5000505559
Encompass
This wire-backed pleated air filter is designed for effective particle capture in various applications. Featuring a MERV 10 rating and 55% efficiency, it is constructed from synthetic media with a durable paperboard frame.
  • Wire-backed pleated design for enhanced stability
  • MERV 10 rating ensures efficient particle filtration
  • Synthetic media offers reliable performance
  • Paperboard frame provides structural integrity
  • Nominal size of 18 in high, 25 in wide, and 2 in deep
  • Capable of handling 1550 CFM air flow at 500 FPM
  • Meets ASHRAE 52.2-2012 and UL 900 standards
  • Suitable for use with any unit
Catalog No. 50-005-05559 Supplier MSC Supplier No. 08587479
This wire-backed pleated air filter is designed for effective particle capture in various applications. Featuring a MERV 10 rating and 55% efficiency, it is constructed from synthetic media with a durable paperboard frame.
  • Wire-backed pleated design for enhanced stability
  • MERV 10 rating ensures efficient particle filtration
  • Synthetic media offers reliable performance
  • Paperboard frame provides structural integrity
  • Nominal size of 18 in high, 25 in wide, and 2 in deep
  • Capable of handling 1550 CFM air flow at 500 FPM
  • Meets ASHRAE 52.2-2012 and UL 900 standards
  • Suitable for use with any unit