Learn More
Med Vet International Sterile 3-Ply Surgical Drape, 18-in x 26-in, with 2.75-in (6.9cm) dia. fenestration, 50/Box
Catalog No. NC9047621
Encompass_Preferred
Excellent for use during minor surgical procedures. Two layers of absorbent tissue with a polyethylene lining in between help eliminate risk of bacterial migration and moisture strike through. Latex-free.
Catalog No. NC9047621 Supplier Med Vet International Supplier No. 20002
Excellent for use during minor surgical procedures. Two layers of absorbent tissue with a polyethylene lining in between help eliminate risk of bacterial migration and moisture strike through. Latex-free.