Learn More
Cayman Chemical 19 20-EDP EthnolamIde 100ug
SDP
Catalog No. 502935801
Encompass_Preferred
An ω-3 endocannabinoid epoxide and CB receptor agonist (EC50s = 108 and 280 nM for CB1 and CB2, respectively); reduces the viability of 143B metastatic osteosarcoma cells at 25 µM; decreases the production of IL-6 and increases LPS-induced production of IL-10 in BV-2 microglia at 2.5-10 µM; decreases LPS-induced cytotoxicity in BV-2 microglia at 5-10 µM; decreases nitrite production at 7.5 µM, an effect that can be partially reversed by AM630 and GW 9662; induces vasodilation of isolated preconstricted bovine coronary arteries (ED50 = 1.9 µM); tube formation by HMVECs
Catalog No. 50-293-5801 Supplier Cayman Chemical Supplier No. 25917100 UG
An ω-3 endocannabinoid epoxide and CB receptor agonist (EC50s = 108 and 280 nM for CB1 and CB2, respectively); reduces the viability of 143B metastatic osteosarcoma cells at 25 µM; decreases the production of IL-6 and increases LPS-induced production of IL-10 in BV-2 microglia at 2.5-10 µM; decreases LPS-induced cytotoxicity in BV-2 microglia at 5-10 µM; decreases nitrite production at 7.5 µM, an effect that can be partially reversed by AM630 and GW 9662; induces vasodilation of isolated preconstricted bovine coronary arteries (ED50 = 1.9 µM); tube formation by HMVECs