Learn More
Cayman Chemical 19 20-EpDTE 100ug
SDP
Catalog No. 502985294
Encompass_Preferred
An oxylipin; an oxidative metabolite of DPA; formed via CYP metabolism of DPA; can be further metabolized to (±)19(20)-DiHDTE by epoxide hydrolase
Catalog No. 50-298-5294 Supplier Cayman Chemical Supplier No. 20331100 UG
An oxylipin; an oxidative metabolite of DPA; formed via CYP metabolism of DPA; can be further metabolized to (±)19(20)-DiHDTE by epoxide hydrolase