Learn More
Cayman Chemical 19 20EpDPA Fatty Acids and Der
SDP
Catalog No. 502889064
Encompass_Preferred
An epoxide metabolite of DHA; formed from DHA by various CYP isoforms; suppresses hCLS formation and liver fibrosis in wild-type mice in a model of NASH at 100 ng/animal, i.p., intraplantar administration induces acute mechanical hyperalgesia in mice
Catalog No. 50-288-9064 Supplier Cayman Chemical Supplier No. 10175100 UG
An epoxide metabolite of DHA; formed from DHA by various CYP isoforms; suppresses hCLS formation and liver fibrosis in wild-type mice in a model of NASH at 100 ng/animal, i.p., intraplantar administration induces acute mechanical hyperalgesia in mice