Learn More
Cayman Chemical 19 20EpDPA Fatty Acids and Der
Supplier: Cayman Chemical 10175500 UG
An epoxide metabolite of DHA; formed from DHA by various CYP isoforms; suppresses hCLS formation and liver fibrosis in wild-type mice in a model of NASH at 100 ng/animal, i.p., intraplantar administration induces acute mechanical hyperalgesia in mice