Learn More
Medchemexpress LLC 19,21-Dinorchol-4-en-20(22)-yn-3-one,17-hydroxy-23,23-dimethyl-11-[4-[methyl(1-methylethyl)amino]phe | 2222344-98-9 | 98.1% | 501.74 | 1 MG
SDP
Catalog No. 5000438547
Encompass_Preferred
ORIC-101 is a highly potent and selective glucocorticoid receptor antagonist, with an EC50 of 5.6 nM. It has anti-cancer activity. Additionally, ORIC-101 is a click chemistry reagent, which means it contains an Alkyne group and can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Highly potent and selective glucocorticoid receptor antagonist
  • Anti-cancer activity
  • Click chemistry reagent
  • Contains an Alkyne group
  • Can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups
Catalog No. 50-004-38547 Supplier Medchemexpress LLC Supplier No. HY1127101MG
ORIC-101 is a highly potent and selective glucocorticoid receptor antagonist, with an EC50 of 5.6 nM. It has anti-cancer activity. Additionally, ORIC-101 is a click chemistry reagent, which means it contains an Alkyne group and can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Highly potent and selective glucocorticoid receptor antagonist
  • Anti-cancer activity
  • Click chemistry reagent
  • Contains an Alkyne group
  • Can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups