Learn More
Ace Glass, Inc. 19/22 170mm distillation column
SDP
Catalog No. 50976750
Encompass_Preferred
19/22 170mm long Vigreux distillation column. Length in millimeters refers to the effective length as measured from the lowest to the highest indent of the column.
Catalog No. 50-976-750 Supplier Ace Glass, Inc. Supplier No. 934513
19/22 170mm long Vigreux distillation column. Length in millimeters refers to the effective length as measured from the lowest to the highest indent of the column.