Learn More
Ace Glass, Inc. PTFE adapter bellows, 19/22 outer top, 19/22 inner bottom, 90mm length
SDP
Catalog No. 50977983
Encompass
90mm PTFE bellows adapter with 19/22 outer top and inner bottom joints. Bellows is used for correct alignment of standard taper joints and relieves stress in reaction systems. Bellows operates to 200 degrees C.
Catalog No. 50-977-983 Supplier Ace Glass, Inc. Supplier No. 1344119
90mm PTFE bellows adapter with 19/22 outer top and inner bottom joints. Bellows is used for correct alignment of standard taper joints and relieves stress in reaction systems. Bellows operates to 200 degrees C.