Learn More
Ace Glass, Inc. Complete 19/22 6mm bearing, straight adapter style
SDP
Catalog No. 50975967
Encompass_Preferred
19/22 6mm straight-adapter style bearing includes all parts needed. Self-aligning, lubricant free PTFE Trubore bearing is designed for use with 6mm O.D. stainless steel or polished glass shafts.
Catalog No. 50-975-967 Supplier Ace Glass, Inc. Supplier No. 8066120
19/22 6mm straight-adapter style bearing includes all parts needed. Self-aligning, lubricant free PTFE Trubore bearing is designed for use with 6mm O.D. stainless steel or polished glass shafts.