Learn More
Ace Glass, Inc. Adapter, 19/22, 5/16in size A hose connection, #11 Ace-Thred complete with 7506-02 nylon bushing and 7855-708 O-ring
SDP
Supplier:  Ace Glass, Inc. 526136
Encompass_Preferred
Adapter with a 19/22 joint at the bottom, 5/16 size A serrated hose connection, and a #11 Ace-Thred at top. Complete with nylon bushing and O-ring.
Catalog No. 50-978-339