Learn More
Ace Glass, Inc. Adapter, 19/22, 5/16in size A hose connection, #7 Ace-Thred, complete with 5029-10 nylon bushing and 7855-704 O-ring
SDP
Catalog No. 50971796
Encompass_Preferred
Adapter with a 19/22 joint at the bottom, 5/16 size A serrated hose connection, and a #7 Ace-Thred at top. Complete with nylon bushing and O-ring.
Catalog No. 50-971-796 Supplier Ace Glass, Inc. Supplier No. 526112
Adapter with a 19/22 joint at the bottom, 5/16 size A serrated hose connection, and a #7 Ace-Thred at top. Complete with nylon bushing and O-ring.