Learn More
Ace Glass, Inc. 19/22 vacuum take-off adapter, stem even with joint bottom
SDP
Supplier:  Ace Glass, Inc. 912407
Encompass_Preferred
19/22 vacuum take-off adapter with the outer joint at an angle of 105 degrees. Stem is even with the bottom joint. Hose connection is opposite the top joint.
Catalog No. 50-979-974