Learn More
Chemglass Life Sciences 19/22 Inner Uncut Drip Joint
SDP
Catalog No. 501531903
Encompass_Preferred
19/22 Inner Uncut Drip Joint. Full or medium length standard taper inner joint having an uncut drip tip. Tooled on the same tubing sizes as the CG-100 inner joints.
Catalog No. 50-153-1903 Supplier Chemglass Life Sciences Supplier No. CGB11703
19/22 Inner Uncut Drip Joint. Full or medium length standard taper inner joint having an uncut drip tip. Tooled on the same tubing sizes as the CG-100 inner joints.