Learn More
Chemglass Life Sciences Drying Tube, 75° Degree, 19/22 Inner Joint, # 00 Rubber Stopper
SDP
Catalog No. CG129519
Encompass_Preferred
Drying Tube, 75¿ Degree, 19/22 Inner Joint, # 00 Rubber Stopper. The 75¿ angle drying tube is for use in fume hoods where space is at a minimum. Supplied complete with rubber stopper, glass inlet tube, and a lower inner joint.
Catalog No. CG129519 Supplier Chemglass Life Sciences Supplier No. CG129519
Drying Tube, 75¿ Degree, 19/22 Inner Joint, # 00 Rubber Stopper. The 75¿ angle drying tube is for use in fume hoods where space is at a minimum. Supplied complete with rubber stopper, glass inlet tube, and a lower inner joint.