Learn More
Chemglass Life Sciences 19/22 Inner Joint with Hooks, on 17mm OD x 1.8mm W tubing
SDP
Catalog No. CG10702
Encompass_Preferred
Inner joint with two hooks 180º apart just above the grind. 19/22 Inner Joint with Hooks, on 17mm OD x 1.8mm W tubing.
Catalog No. CG10702 Supplier Chemglass Life Sciences Supplier No. CG10702
Inner joint with two hooks 180º apart just above the grind. 19/22 Inner Joint with Hooks, on 17mm OD x 1.8mm W tubing.