Learn More
Chemglass Life Sciences 19/22 Outer Joint, Dewar Seal, 40mm OD Flare
SDP
Catalog No. CG13133
Encompass_Preferred
19/22 Outer Joint, Dewar Seal, 40mm OD Flare. Used in construction of apparatus requiring the outer member to be jacketed. Configuration of flare permits seal to be made without having to regrind the joint.
Catalog No. CG13133 Supplier Chemglass Life Sciences Supplier No. CG13133
19/22 Outer Joint, Dewar Seal, 40mm OD Flare. Used in construction of apparatus requiring the outer member to be jacketed. Configuration of flare permits seal to be made without having to regrind the joint.