Learn More
Chemglass Life Sciences 19/22 Outer Joint, Medium Length, on 22mm OD x 1.8mm W tubing
SDP
Catalog No. CG10507
Encompass_Preferred
Ground Joint, Outer, Medium Length. 19/22 Outer Joint, Medium Length, on 22mm OD x 1.8mm W tubing.
Catalog No. CG10507 Supplier Chemglass Life Sciences Supplier No. CG10507
Ground Joint, Outer, Medium Length. 19/22 Outer Joint, Medium Length, on 22mm OD x 1.8mm W tubing.