Learn More
Chemglass Life Sciences 19/22 Outer Joint, Medium Length, on 17mm OD x 1.8mm W tubing
SDP
Catalog No. CG10606
Encompass_Preferred
19/22 Outer Joint, Medium Length, on 17mm OD x 1.8mm W tubing. Standard taper outer joint tooled on the same diameter tubing as the CG-104 inner joints.
Catalog No. CG10606 Supplier Chemglass Life Sciences Supplier No. CG10606
19/22 Outer Joint, Medium Length, on 17mm OD x 1.8mm W tubing. Standard taper outer joint tooled on the same diameter tubing as the CG-104 inner joints.