Learn More
Chemglass Life Sciences 50mm Powder Funnel, 19/22 Inner Joint
SDP
Catalog No. CG172011
Encompass_Preferred
50mm Powder Funnel, 19/22 Inner Joint. Useful for the introduction of solids or liquids into reactions under atmospheric conditions. Funnels have 60º side angle and a standard taper inner joint.
Catalog No. CG172011 Supplier Chemglass Life Sciences Supplier No. CG172011
50mm Powder Funnel, 19/22 Inner Joint. Useful for the introduction of solids or liquids into reactions under atmospheric conditions. Funnels have 60º side angle and a standard taper inner joint.