Learn More
Chemglass Life Sciences Septum Stopper, Sleeve Type, For 19/22-19/38 Outer Joints, Red
SDP
Catalog No. CG302296
Encompass_Preferred
Septum Stopper, Sleeve Type, For 19/22-19/38 Outer Joints, Red. Sleeve type stoppers with a hollow plugs and sleeve extensions that folds down over the neck of a vessel, tubing, etc. The diaphragm can be punctured with a syringe needle, re-sealing after the needle is withdrawn.
Catalog No. CG302296 Supplier Chemglass Life Sciences Supplier No. CG302296
Septum Stopper, Sleeve Type, For 19/22-19/38 Outer Joints, Red. Sleeve type stoppers with a hollow plugs and sleeve extensions that folds down over the neck of a vessel, tubing, etc. The diaphragm can be punctured with a syringe needle, re-sealing after the needle is withdrawn.