Learn More
Ace Glass, Inc. Connecting adapter with 19/38 top female and 14/20 bottom male standard taper joints.
SDP
Catalog No. 50979964
Encompass_Preferred
Connecting adapter with 19/38 top female and 14/20 bottom male standard taper joints.
Catalog No. 50-979-964 Supplier Ace Glass, Inc. Supplier No. 909221
Connecting adapter with 19/38 top female and 14/20 bottom male standard taper joints.