Learn More
Ace Glass, Inc. Connecting adapter, enlarging, 19/38 outer top, 14/35inner bottom
SDP
Supplier:  Ace Glass, Inc. 500506
Encompass_Preferred
Connecting, enlarging adapter featuring a 19/38 outer top female joint and a 14/35 inner bottom male joint.
Catalog No. 50-978-270