Learn More
Ace Glass, Inc. PTFE adapter bellows. 19/38 outer top, 19/38 inner bottom120 mm length
SDP
Catalog No. 50977984
Encompass
120mm PTFE bellows adapter with 19/38 outer top and inner bottom joints. Bellows is used for correct alignment of standard taper joints and relieves stress in reaction systems. Bellows operates to 200 degrees C.
Catalog No. 50-977-984 Supplier Ace Glass, Inc. Supplier No. 1344123
120mm PTFE bellows adapter with 19/38 outer top and inner bottom joints. Bellows is used for correct alignment of standard taper joints and relieves stress in reaction systems. Bellows operates to 200 degrees C.