Learn More
Chemglass Life Sciences 19/38 Side Seal Joints, Sidearm OD: 10.0mm (2), Hole Size: 4mm
SDP
Catalog No. CG13203
Encompass_Preferred
19/38 Side Seal Joints, Sidearm OD: 10.0mm (2), Hole Size: 4mm. Outer joint having a tubulation sealed at the midpoint of the ground portion, and a corresponding hole drilled in the inner joint permitting venting of the side tube when the inner joint is turned to the proper position. The 19/38 sizes are compatible with Buchi Rotary Evaporators. Part number CG-132-03 has two sidearms sealed 180º apart and is used for replacement on the vertical cold trap condenser assembly.
Catalog No. CG13203 Supplier Chemglass Life Sciences Supplier No. CG13203
19/38 Side Seal Joints, Sidearm OD: 10.0mm (2), Hole Size: 4mm. Outer joint having a tubulation sealed at the midpoint of the ground portion, and a corresponding hole drilled in the inner joint permitting venting of the side tube when the inner joint is turned to the proper position. The 19/38 sizes are compatible with Buchi Rotary Evaporators. Part number CG-132-03 has two sidearms sealed 180º apart and is used for replacement on the vertical cold trap condenser assembly.