Learn More
Chemglass Life Sciences 19/38 Outer Joint, 22mm X 1.8W Tubing
SDP
Catalog No. 501531890
Encompass_Preferred
19/38 Outer Joint, 22mm X 1.8W Tubing. Standard taper outer joint tooled on tubing approximately the same diameter as the top reinforcing ring, affording no hold-up below joint.
Catalog No. 50-153-1890 Supplier Chemglass Life Sciences Supplier No. CGB10105
19/38 Outer Joint, 22mm X 1.8W Tubing. Standard taper outer joint tooled on tubing approximately the same diameter as the top reinforcing ring, affording no hold-up below joint.