Learn More
Chemglass Life Sciences 19/38 Side Seal Joints, Sidearm OD: 10.0mm, Hole Size: 4mm
SDP
Catalog No. CG13202
Encompass_Preferred
19/38 Side Seal Joints, Sidearm OD: 10.0mm, Hole Size: 4mm. Outer joint having a tubulation sealed at the midpoint of the ground portion, and a corresponding hole drilled in the inner joint permitting venting of the side tube when the inner joint is turned to the proper position.The 19/38 sizes are compatible with Buchi Rotary Evaporators. Part number CG-132-02 has a single sidearm and a drip tip permitting the seal to be made for the PTFE feed tube.
Catalog No. CG13202 Supplier Chemglass Life Sciences Supplier No. CG13202
19/38 Side Seal Joints, Sidearm OD: 10.0mm, Hole Size: 4mm. Outer joint having a tubulation sealed at the midpoint of the ground portion, and a corresponding hole drilled in the inner joint permitting venting of the side tube when the inner joint is turned to the proper position.The 19/38 sizes are compatible with Buchi Rotary Evaporators. Part number CG-132-02 has a single sidearm and a drip tip permitting the seal to be made for the PTFE feed tube.