Learn More
PHCbi 7.6 cu.ft. Pharmaceutical Refrigerator
Catalog No. 502740418
Encompass_Preferred
PHCbi brand pharmaceutical refrigerator units are engineered from the inside out to create, maintain, and restore precise temperatures necessary for vaccine storage. Designed specifically for storage not only of vaccines but also other pharmaceuticals, our purpose-built refrigerator units are designed for temperature uniformity and stability that is not commonly found in household or commercial cabinets. PHCbi brand vaccine cold chain storage solutions satisfy current and evolving storage protocols essential for continued vaccine efficacy. 4 Upper, 1 Lower (allowable load 44 lbs. (20 kg)/shelf; inner dimensions upper W x D: 12.8" x 13.3" (326 x 339 mm), inner dimensions lower W x D: 11.8" x 11.8" (300 x 300 mm))
Catalog No. 50-274-0418 Supplier PHCbi Supplier No. MPRN200HPA
PHCbi brand pharmaceutical refrigerator units are engineered from the inside out to create, maintain, and restore precise temperatures necessary for vaccine storage. Designed specifically for storage not only of vaccines but also other pharmaceuticals, our purpose-built refrigerator units are designed for temperature uniformity and stability that is not commonly found in household or commercial cabinets. PHCbi brand vaccine cold chain storage solutions satisfy current and evolving storage protocols essential for continued vaccine efficacy. 4 Upper, 1 Lower (allowable load 44 lbs. (20 kg)/shelf; inner dimensions upper W x D: 12.8" x 13.3" (326 x 339 mm), inner dimensions lower W x D: 11.8" x 11.8" (300 x 300 mm))