Learn More
MSC 3M 7000121418 24 oz Aerosol Clear Spray Adhesive High Tack, 200°F Heat Resistance, 50 Sq Ft Coverage, High Strength Bond, 30 min Max Bonding Time, Flammable, Series High-Strength 90
Catalog No. 501296899
Encompass
3M 7000121418 24 oz Aerosol Clear Spray Adhesive High Tack, 200°F Heat Resistance, 50 Sq Ft Coverage, High Strength Bond, 30 min Max Bonding Time, Flammable, Series High-Strength 90
Catalog No. 50-129-6899 Supplier MSC Supplier No. 54137294
3M 7000121418 24 oz Aerosol Clear Spray Adhesive High Tack, 200°F Heat Resistance, 50 Sq Ft Coverage, High Strength Bond, 30 min Max Bonding Time, Flammable, Series High-Strength 90