Learn More
Cayman Chemical 19-alkyne Cholesterol, Purity: ≥95%, CAS: 2058319-71-2, 10 mg
SDP
Catalog No. 502971344
Encompass_Preferred
An alkyne derivative of cholesterol; has been used to track cholesterol localization and subcellular distribution in NIH3T3 cells and to study the role of cholesterol in Hh signal transduction in Shh-LIGHT2 cells using click chemistry and various reporter azides
Catalog No. 50-297-1344 Supplier Cayman Chemical Supplier No. 1644610 MG
An alkyne derivative of cholesterol; has been used to track cholesterol localization and subcellular distribution in NIH3T3 cells and to study the role of cholesterol in Hh signal transduction in Shh-LIGHT2 cells using click chemistry and various reporter azides