Learn More
Perkin Elmer US LLC 19 ga. Custodion CME Sampling Device with 3 Replacement Coils
Catalog No. 502104889
Encompass_Preferred
Sampling just got easier - Simplify your collection devices to save time obtaining field samples by investing in CME technology. CME is ideal for liquids including explosives, accelerants, drugs of abuse and, semi-volatiles. This novel technique consisting of a finely coiled wire that traps liquid samples by capillary action. When used with the Torion T-9 Portable GC/MS, in-field sample analysis is quick, easy, and reliable.
Catalog No. 50-210-4889 Supplier Perkin Elmer US LLC Supplier No. NTSC19SN3B191
Sampling just got easier - Simplify your collection devices to save time obtaining field samples by investing in CME technology. CME is ideal for liquids including explosives, accelerants, drugs of abuse and, semi-volatiles. This novel technique consisting of a finely coiled wire that traps liquid samples by capillary action. When used with the Torion T-9 Portable GC/MS, in-field sample analysis is quick, easy, and reliable.