Learn More
Justrite™ 19 Gallon Under Fume Hood Solvent/Flammable Liquid Safety Cabinet With Manual-Closing Doors
GSA_VAGREENER_CHOICE
Catalog No. 19822334 Shop All Justrite Safety Group Products
Change view
Click to view available options
Color:
Light Neutral
Silver
Yellow
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-822-334 Light Neutral
19-822-335 Silver
19-822-336 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-822-334 Supplier Justrite™ Supplier No. 883007
  • $3,620.00
    $2,771.65 / Each of 1
    Save $848.35
    23% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Increase workplace safety and improve cleaniliness with the proper safety and storage equipment. The Justrite™ 19 Gallon Under Fume Hood Solvent/Flammable Liquid Safety Cabinet With Manual-Closing Doors is fully compliant and designed for safe storage of solvents and flammables in laboratories.
  • Justrite's Biohazard Waste Containers meet OSHA 29 CFR 1910.1030 for the segregation of infectious waste
  • Use them to store contaminated laundry as well as other regulated waste; however, not for the storage of sharps
  • Sturdy steel cans feature leakproof construction with foot-operated, self-closing lid
  • Simple, foot pedal operation opens the cover automatically to safely dispose of biohazard waste
  • Upon release of the pedal, the can cover closes quickly to Reduce the risk of contamination
  • A biohazard bag is held securely around the circumference of the can top
  • Optional label kit #25880 complies with California Health and Safety Code 118280 requiring warning labels to be visible from all sides of the container
  • Kit includes three labels and instructions
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Door Style Manual-close
Load Bearing Capacity 19 gal.
Color Light Neutral