Learn More
Medchemexpress LLC 19-Hydroxy cholester 1mg | 561-63-7 | 1 MG
SDP
Catalog No. 5000227251
Encompass_Preferred
19-Hydroxy cholesterol is a hydroxylated cholesterol metabolite supplied as a powdered biochemical reagent for research applications. It has CAS 561-63-7, chemical formula C27H46O2, and molecular weight 402.65. Typical purity is ≥99.0% (HPLC). Store frozen to preserve stability.
  • High purity (≥99.0% by HPLC).
  • Supplied as a stable powder for easy handling.
  • Suitable as a reference standard in biochemical assays.
  • Long-term storage at -20°C preserves integrity.
  • Soluble in common organic solvents for assay preparation.
Catalog No. 50-002-27251 Supplier Medchemexpress LLC Supplier No. HY1146491MG
19-Hydroxy cholesterol is a hydroxylated cholesterol metabolite supplied as a powdered biochemical reagent for research applications. It has CAS 561-63-7, chemical formula C27H46O2, and molecular weight 402.65. Typical purity is ≥99.0% (HPLC). Store frozen to preserve stability.
  • High purity (≥99.0% by HPLC).
  • Supplied as a stable powder for easy handling.
  • Suitable as a reference standard in biochemical assays.
  • Long-term storage at -20°C preserves integrity.
  • Soluble in common organic solvents for assay preparation.